Boss Head, Double Swivel
470137-842
- Descripción:Boss Head - Double Swivel
- Referencia:470137-842
- Nº de proveedor:CH0666
Our swiveling boss head allows you to hold two rods at any angle.
Made of die cast zinc with the ability to hold rods up to 16 mm in diameter.