Orders LinkContactUsLinkComponent
Boss Head, Double Large
Boss Head, Double Large
Boss Head Double Large, Chrome Plated
  470137-838
Supplier #:  CH0665A
Boss Head, Double Large
  470137-838
Boss Head, Double Large
Boss Head, Double Large
Boss Head Double Large, Chrome Plated
$414.68
Each (1UN)
Lab Aid, Many Uses
This versatile boss head can hold rods ranging from 5-20mm in diameter. Choose from chrome-plated or black finish. The die cast boss head is fitted with plastic cap screws for easy tightening.
  • Description:
    Boss Head Double Large, Chrome Plated