Universal Clamp Holder, Jumbo
Jumbo Universal Clamp Holder, 20 mm
Catalog # 470137-836
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: CH0664JM
Jump to:
Product Details & Documents
Large Clamp Capacity You Need.
- Accommodates rods and flat boards
This larger clamp holder accommodates rods up to 20mm in diameter and flat boards up to 18mm thick.
Especificaciones
- Description:Jumbo Universal Clamp Holder
- Rod Diameter:20 mm