Drying Basket, Polypropylene
Drying Basket, Polypropylene
Catalog # 470137-796
Supplier: VWR International
Jump to:
Product Details & Documents
Dry Lab Equipment Safely
This large polypropylene drying basket stands up to repeated autoclaving and has perforations for draining various pieces of lab equipment. Size: 15 3/4"L x 15 3/4"W x 4"H.
Especificaciones
- Description:Drying Basket, Polypropylene