Ring and Disk Moment of Inertia Apparatus
Catalog # 470137-746
Supplier: AMERICAN SCIENTIFIC LLC SE
Small Business Enterprise
Specifications
- Descripción:Ring and Disk Moment of Inertia Apparatus
- Dimensiones:15 dia. x 5 cm thick
- Referencia:470137-746
- Nº de proveedor:3479-00
Specifications
About this item
Easy to Study Inertia and Mass
Use this handy apparatus to help students study the dependence of moment of inertia on the distribution of mass. It's easy to see differences in acceleration when the metal ring and wooden disk of the same mass are rolled on the provided inclined plane.