Manver 2 Hardness Indicator Powder Pillows, Hach
Catalog # 89230-886
Supplier: HACH CO
Product available on GSA Advantage®
CAS Number:
Specifications
- Método químico:EDTA
- Descripción:Manver 2 Hardness Indicator Powder Pillows
- Tamaño:50 mL Sample
- Referencia:89230-886
- Nº de proveedor:85199
- No. of Tests:100
Specifications
About this item
Indicator for determination of total hardness and chelants by titration. Powder Pillows for 50 mL samples.
Powder Pillows for 50 mL samples.
Ordering information: Pack of 100 powder pillows.