qScript™ XLT One-Step RT-qPCR ToughMix®, QuantaBio
Acerca de este artículo
qScript XLT One-Step RT-qPCR ToughMix is a ready-to-use, highly sensitive master mix for reverse transcription quantitative PCR (RT-qPCR) of RNA templates using hybridization probe detection chemistries such as TaqMan® 5’-hydrolysis probes on real-time PCR systems that do not require a passive reference dye. First-strand cDNA synthesis and PCR amplification are carried out in the same tube without opening between procedures. It is ideal for highly sensitive quantification of RNA viruses or low abundance RNA targets as well as high throughput gene-expression studies. The system has been optimized to deliver maximum RT-PCR efficiency, sensitivity, and specificity in reduced reaction volumes and fast cycle times. qScript XLT One-Step RT-qPCR ToughMix contains all required components for RT-qPCR except RNA template and probe. It is compatible with all dual-labeled probe chemistries.
- Tough Tested – Overcomes common inhibitors including polysacharides, heme/hemoglobin, humic acid, melanin.
- Flexible - Use fast or standard qPCR cycling conditions.
- Broad dynamic range- Reliable data from your precious samples every time.
- Multiplexing enabled – Supports detection of up to four targets.
qScript XLT is an engineered M-MLV reverse transcriptase with reduced RNase H activity and improved activity and stability at higher temperatures. The use of higher temperatures (50 to 55°C) for the first-strand step of one-step RT-qPCR provides higher specificity for primer annealing and disruption of RNA secondary structure that can interfere with cDNA synthesis. qScript XLT One-Step RT-qPCR ToughMix is highly resistant to PCR inhibitors. A key component of the ToughMix is an ultra pure, highly processive thermostable DNA polymerase that is combined with high avidity monoclonal antibodies. This provides an extremely stringent automatic hot-start that minimizes the potential for primer-dimer and other non-specific PCR artifacts. The light blue color of the AccuVue™ tracer dye simplifies reaction assembly in white, or clear, plates and helps to minimize pipetting or mixing errors. It does not interfere with qPCR performance or affect the stability of the product.
Different real-time PCR systems employ different strategies for the normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagent to your specific instrument. The qScript XLT One-Step RT-qPCR Kit does not contain a passive reference dye.
Real-Time PCR Instrument Compatibility: Roche LightCycler™ 480; Qiagen/Corbett Rotor-Gene® Q; Qiagen/Corbett Rotor-Gene® 6000; Qiagen/Corbett Rotor-Gene® 3000; Eppendorf Mastercycler® ep realplex; Eppendorf Mastercycler® ep realplex2 s; Cepheid SmartCycler®; Bio-Rad CFX96™; Bio-Rad CFX384™; Bio-Rad/MJ Opticon®; Bio-Rad/MJ Opticon 2; Bio-Rad/MJ Chromo4™; Bio-Rad MiniOpticon™; Illumina Eco qPCR; Thermo Scientific PikoReal Cycler; BioRad iCycler iQ™; BioRad MyiQ™; BioRad iQ™5.
Ordering information: Contains Reagent Description; qScript XLT One-Step RT-qPCR ToughMix (2X); 2X reaction buffer containing dATP, dCTP, dGTP, dTTP, magnesium chloride, qScript XLT reverse transcriptase, RNase inhibitor protein, hot-start DNA polymerase, AccuVue blue qPCR dye, and stabilizers.
Packaging: Stable for one year when stored in a constant temperature freezer at -20°C protected from light. Repeated freezing and thawing does not affect RT-qPCR performance.
Caution: For research use only. Not intended for any animal or human therapeutic or diagnostic use. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
6 Options Available Below
- El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.
- Return Policy
Product Details & Documents
qScript XLT One-Step RT-qPCR ToughMix is a ready-to-use, highly sensitive master mix for reverse transcription quantitative PCR (RT-qPCR) of RNA templates using hybridization probe detection chemistries such as TaqMan® 5’-hydrolysis probes on real-time PCR systems that do not require a passive reference dye. First-strand cDNA synthesis and PCR amplification are carried out in the same tube without opening between procedures. It is ideal for highly sensitive quantification of RNA viruses or low abundance RNA targets as well as high throughput gene-expression studies. The system has been optimized to deliver maximum RT-PCR efficiency, sensitivity, and specificity in reduced reaction volumes and fast cycle times. qScript XLT One-Step RT-qPCR ToughMix contains all required components for RT-qPCR except RNA template and probe. It is compatible with all dual-labeled probe chemistries.
- Tough Tested – Overcomes common inhibitors including polysacharides, heme/hemoglobin, humic acid, melanin.
- Flexible - Use fast or standard qPCR cycling conditions.
- Broad dynamic range- Reliable data from your precious samples every time.
- Multiplexing enabled – Supports detection of up to four targets.
qScript XLT is an engineered M-MLV reverse transcriptase with reduced RNase H activity and improved activity and stability at higher temperatures. The use of higher temperatures (50 to 55°C) for the first-strand step of one-step RT-qPCR provides higher specificity for primer annealing and disruption of RNA secondary structure that can interfere with cDNA synthesis. qScript XLT One-Step RT-qPCR ToughMix is highly resistant to PCR inhibitors. A key component of the ToughMix is an ultra pure, highly processive thermostable DNA polymerase that is combined with high avidity monoclonal antibodies. This provides an extremely stringent automatic hot-start that minimizes the potential for primer-dimer and other non-specific PCR artifacts. The light blue color of the AccuVue™ tracer dye simplifies reaction assembly in white, or clear, plates and helps to minimize pipetting or mixing errors. It does not interfere with qPCR performance or affect the stability of the product.
Different real-time PCR systems employ different strategies for the normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagent to your specific instrument. The qScript XLT One-Step RT-qPCR Kit does not contain a passive reference dye.
Real-Time PCR Instrument Compatibility: Roche LightCycler™ 480; Qiagen/Corbett Rotor-Gene® Q; Qiagen/Corbett Rotor-Gene® 6000; Qiagen/Corbett Rotor-Gene® 3000; Eppendorf Mastercycler® ep realplex; Eppendorf Mastercycler® ep realplex2 s; Cepheid SmartCycler®; Bio-Rad CFX96™; Bio-Rad CFX384™; Bio-Rad/MJ Opticon®; Bio-Rad/MJ Opticon 2; Bio-Rad/MJ Chromo4™; Bio-Rad MiniOpticon™; Illumina Eco qPCR; Thermo Scientific PikoReal Cycler; BioRad iCycler iQ™; BioRad MyiQ™; BioRad iQ™5.
Ordering information: Contains Reagent Description; qScript XLT One-Step RT-qPCR ToughMix (2X); 2X reaction buffer containing dATP, dCTP, dGTP, dTTP, magnesium chloride, qScript XLT reverse transcriptase, RNase inhibitor protein, hot-start DNA polymerase, AccuVue blue qPCR dye, and stabilizers.
Packaging: Stable for one year when stored in a constant temperature freezer at -20°C protected from light. Repeated freezing and thawing does not affect RT-qPCR performance.
Caution: For research use only. Not intended for any animal or human therapeutic or diagnostic use. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Documents
Certificates
Buscar por número de loteAzure Biosystems Cielo Products.pdf
Promotion Material
One-Step SYBR Green.pdf
Datasheet
VWR CDC COVID-19 RUO RT-qPCR Protocol
Product Literature
Specifications for Product Family
Especificaciones
Especificaciones
- Catalog No:
- 89236-666
- 89236-668
- 89236-670
- 89236-672
- 89236-674
- 89236-676
- Application:
- RT-qPCR probe
- RT-qPCR probe
- RT-qPCR probe
- RT-qPCR probe
- RT-qPCR probe
- RT-qPCR probe
- One step:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Multiplexing:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Inhibitor-rich sample:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
Especificaciones
Product Family Options
One-Step RT-qPCR ToughMix®
Información del producto
- Cycler compatibilityNo. of ReactionsAvailabilityPrice
- 89236-666qScript™ XLT One-Step RT-qPCR ToughMix®qScript™ XLT One-Step RT-qPCR ToughMix®Agilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480100× 20 µlLog in Para disponibilidadEach (1UN)$6,146.26Supplier Number: 95132-100El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.Especificaciones:
More
Specifications
Cat. No.89236-666ApplicationRT-qPCR probeOne stepYesMultiplexingYesInhibitor-rich sampleYes - 89236-668qScript™ XLT One-Step RT-qPCR ToughMix®qScript™ XLT One-Step RT-qPCR ToughMix®Agilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480500 (20 µl)Log in Para disponibilidadEach (1UN)$21,379.93Supplier Number: 95132-500El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.Especificaciones:
More
Specifications
Cat. No.89236-668ApplicationRT-qPCR probeOne stepYesMultiplexingYesInhibitor-rich sampleYes
One-Step RT-qPCR ToughMix®, ROX
Información del producto
- Cycler compatibilityNo. of ReactionsAvailabilityPrice
- 89236-670qScript™ XLT One-Step RT-qPCR ToughMix®, ROX™qScript™ XLT One-Step RT-qPCR ToughMix®, ROX™ABI 7900HT/7900HT Fast/StepOnePlus™100× 20 µlLog in Para disponibilidadEach (1UN)$6,183.97Supplier Number: 95133-100El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.Especificaciones:
More
Specifications
Cat. No.89236-670ApplicationRT-qPCR probeOne stepYesMultiplexingYesInhibitor-rich sampleYes - 89236-672qScript™ XLT One-Step RT-qPCR ToughMix®, ROX™qScript™ XLT One-Step RT-qPCR ToughMix®, ROX™ABI 7900HT/7900HT Fast/StepOnePlus™500 (20 µl)Log in Para disponibilidadEach (1UN)$21,379.93Supplier Number: 95133-500El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.Especificaciones:
More
Specifications
Cat. No.89236-672ApplicationRT-qPCR probeOne stepYesMultiplexingYesInhibitor-rich sampleYes
One-Step RT-qPCR ToughMix®, Low ROX
Información del producto
- Cycler compatibilityNo. of ReactionsAvailabilityPrice
- 89236-674qScript™ XLT One-Step RT-qPCR ToughMix®, Low ROX™qScript™ XLT One-Step RT-qPCR ToughMix®, Low ROX™ABI 7500/7500 Fast/QuantStudio™, Agilent AriaMx, Douglas Scientific IntelliQube®, Stratagene Mx100× 20 µlLog in Para disponibilidadEach (1UN)$6,183.97Supplier Number: 95134-100El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.Especificaciones:
More
Specifications
Cat. No.89236-674ApplicationRT-qPCR probeOne stepYesMultiplexingYesInhibitor-rich sampleYes - 89236-676qScript™ XLT One-Step RT-qPCR ToughMix®, Low ROX™qScript™ XLT One-Step RT-qPCR ToughMix®, Low ROX™ABI 7500/7500 Fast/QuantStudio™, Agilent AriaMx, Douglas Scientific IntelliQube®, Stratagene Mx500 (20 µl)Log in Para disponibilidadEach (1UN)$21,379.93Supplier Number: 95134-500El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.Especificaciones:
More
Specifications
Cat. No.89236-676ApplicationRT-qPCR probeOne stepYesMultiplexingYesInhibitor-rich sampleYes