ContactUsLinkComponent
Round-Bottom Flask with Beaded Tube Side Well, Ace Glass Incorporated
Round-Bottom Flask with Beaded Tube Side Well, Ace Glass Incorporated
Catálogo # 89056-538
Proveedor:  ACE GLASS, INC.
Product available on GSA Advantage®
Small Business Enterprise
undefined
Round-Bottom Flask with Beaded Tube Side Well, Ace Glass Incorporated
Catálogo # 89056-538
Proveedor:  ACE GLASS, INC.
Número de proveedor:  6935-68
Product available on GSA Advantage®
Small Business Enterprise
Especificaciones
  • [ST] unión No.:
    24/40
  • Capacidad:
    250 ml
  • Referencia:
    89056-538
  • Nº de proveedor:
    6935-68
Especificaciones
Acerca de este artículo
This round-bottom flask has a center neck with [ST] outer joint and a deep beaded side well
The outer joint is tooled with reinforcement rings for added strength and stability. The I.D. of the side well is approximately 9 mm. Fabricated from borosilicate glass.
Información de pedido: For a version of this flask with a side port rather than a deep side well, see 89056-522.