ContactUsLinkComponent
Distillation Apparatus, Methoxy Determination, Ace Glass
Catalog # 89055-556
Supplier:  ACE GLASS, INC.
Product available on GSA Advantage®
Small Business Enterprise
undefined
Distillation Apparatus, Methoxy Determination, Ace Glass
Catalog # 89055-556
Supplier:  ACE GLASS, INC.
Supplier Number:  6549-45
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Descripción:
    Methoxy Determination Distillation Apparatus (Complete)
  • Referencia:
    89055-556
  • Nº de proveedor:
    6549-45
Specifications
About this item
This distillation apparatus is used for methoxy determination as described in U.S
Pharmacopeia, 1995, page 1742. Fabricated from borosilicate glass.
Ordering information: Complete item consists of 20mL boiling flask (89055-550) with a [ST] 14/20 joint and a capillary side arm; scrubber trap column (89055-552) with a [ST] 14/20 inner and [SJ] 35/20 socket; absorption tube (89055-554) with [SJ] 35/20 ball joint and a 2mm bore glass stopcock; and two stainless steel springs (89061-672). Flask and column are supplied with hooks to enable spring connection. Individual components may be purchased separately.