Orders LinkContactUsLinkComponent
Phosphate Buffer, pH 7.0, VWR Chemicals BDH®
±0.1, Phosphate, Poly bottle, 1 L, 7.00, Colorless
Catalog # BDH7447-1
Supplier:   VWR International
Phosphate Buffer, pH 7.0, VWR Chemicals BDH®
Catalog # BDH7447-1
Supplier:  VWR International
CAS Number:  
Phosphate Buffer, pH 7.0, VWR Chemicals BDH®
Phosphate Buffer, pH 7.0, VWR Chemicals BDH®
±0.1, Phosphate, Poly bottle, 1 L, 7.00, Colorless
CAS Number:  
Price
$825.83
Log in Para disponibilidad
Each (1l) Case de 12 (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Suitable for determination of chlorine.
Documents
Especificaciones
Especificaciones
  • Pack type:
    Poly bottle
  • Color:
    Colorless
  • Size:
    1 L
  • pH Buffer Type:
    Phosphate
  • pH:
    7.00
  • pH Accuracy:
    ±0.1
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 