Orders LinkContactUsLinkComponent
VWR® Standard Microplate Vortex Mixers
VWR® Standard Microplate Vortex Mixers
Standard Microplate Vortex Mixer, 120 V, 0.25 A, 30 W, 3.5 mm (0.13")
Catalog # 89399-880
Product available on GSA Advantage®
   Waste Reducing
Supplier:   VWR International
VWR® Standard Microplate Vortex Mixers
Catalog # 89399-880
Supplier:  VWR International
Product available on GSA Advantage®
   Waste Reducing
VWR® Standard Microplate Vortex Mixers
VWR® Standard Microplate Vortex Mixers
Standard Microplate Vortex Mixer, 120 V, 0.25 A, 30 W, 3.5 mm (0.13")
Price
$17,478.52
Log in for availability
Each (1UN)
Product Details & Documents
The VWR® Standard Microplate Vortex Mixer is a variable speed analog mixer that is designed for continuous duty.
  • Designed for continuous duty
  • Designed for shaking microplates or tubes
  • Now with Best-in-Class, Exclusive 5-Year Warranty
The high speed and small orbit is optimal for effectively mixing microplates. Choose from two modes of operation: "Touch" mode which activates mixing when cup head is depressed or "On" mode when using any of the accessory attachments for continuous operation. Analog controls. Continous duty rating.
Certifications: RoHS certified. 120V Unit is CSA certified; 230V Unit is CE marked.
Ordering information: Mixer includes a cup head, single microplate holder, and a 234 cm (92") detachable, 3-wire cord and plug.
Specifications
  • Description:
    Standard Microplate Vortex Mixer
  • Electrical:
    120 V, 0.25 A, 30 W
  • Orbit:
    3.5 mm (0.13")
  • Cat. No.:
    89399-880
  • Capacity:
    2 Microplates
  • Speed range:
    On Mode 300–2500 rpm; Touch Mode 300–3500 rpm
  • Dimensions:
    26.7L x 13.7W x 11.4H cm (10.5 x 5.4 x 4.5")
  • Environmentally Preferable:
    Waste Reducing
  • Weight:
    5.4 kg (12 lbs.)
  • Operating conditions:
    4 to 40°C (39 to 104°F), 20% to 85% relative humidity, non-condensing.
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • VWR® Cup Head
    VWR® Cup Head
    97043-570
    Log in for availability
    Each (1UN) - $1,058.55
Product Information
  • Catalog #
    Availability
    Price
  • VWR® Microplate Holder (Double)
    VWR® Microplate Holder (Double)
    97043-578
    Log in for availability
    Each (1UN) - $1,881.34
Product Information
  • Catalog #
    Availability
    Price
  • VWR® Microplate Holder (Single)
    VWR® Microplate Holder (Single)
    97043-572
    Log in for availability
    Each (1UN) - $1,612.52