Large Pipette Storage Box and Drawer Organizer, PVC, TrippNT
11376-232
: TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Descripción:Large Storage Box
- Referencia:11376-232
- La x An x Al:640x410x80 mm (25x16x3")
- Peso:3 kg (6 lbs.)
- Nº de proveedor:50069
White PVC large pipette storage box and drawer organizer has adjustable compartments and stores and protects supplies and pipettes
The dividers can be moved to accommodate an array of supplies. Made of strong PVC material.