These convenient wipers offer portion control and reduced potential of spillage in and out of the cleanroom.
- Fibers blended by a hydroentanglement procedure that uses water instead of chemicals to bind
- Excellent bidirectional strength and a very low extractable level
Sterile TechniSat® is used for alcohol wiping in sterile environments in pharmaceutical aseptic fill areas, biotechnology manufacturing facilities, microbiological laboratories, sterile suites and prep rooms.
Sterile wipers are saturated with 70% USP grade isopropyl alcohol and packaged in resealable flexpacks. The flexpacks are then double-bagged to maintain sterility. Each package is gamma irradiated to 10 to 6 sterility assurance level. Wipers are lot controlled for traceability.
The blended material of 55% cellulose and 45% polyester provides the high absorbency of a natural fiber with the cleanliness and strength of a synthetic material.