Orders LinkContactUsLinkComponent
VWR® Precision Verification Thermometers
VWR® Precision Verification Thermometers
VWR® Precision Verification Thermometers
Supplier:  VWR International
Certificates
VWR® Precision Verification Thermometers
VWR® Precision Verification Thermometers
4 Options Available Below
Product Details & Documents
To verify sample temperatures in freezers, refrigerators, incubators, or ovens with traceability to NIST. Each item features individual serial numbers and a Statement of Accuracy indicating accuracy traceable to NIST. Thermometer contains organic liquid; bottle contains glass beads.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Insert thermometer into the bottle and place near the samples for best results
  • Thermometers contain organic liquid; bottles are 60ml and contain glass beads
  • Manufactured and tested by our USA quality team; individually serialized
Certifications: Tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST and DKD/PTB.
Ordering information: If an individual Thermometer Calibration Report is required, factory calibrated thermometers or custom calibration services are available. Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
Documents
Product Family Options
Thermometers
Información del producto
  • Range
    Length
    Divisions
    Accuracy
    Availability
    Price
  • 89095-674
    Refrigerator Thermometer
    Refrigerator Thermometer
    ‒2…+10 °C
    180 mm
    0.1 °C
    ±0.4 °C
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,146.71
    $1,146.71
    Log in Para disponibilidad
    Case de 25 (1UN)
    Case de 25 (1UN) - $1,249.91
    $1,249.91
    Each (1UN)
       $1,146.71
    Case de 25 (1UN)
       $1,249.91
  • 89095-694
    Incubator Thermometer
    Incubator Thermometer
    +25...+45 °C
    190 mm
    0.1 °C
    ±0.4 °C
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,094.33
    $1,094.33
    Log in Para disponibilidad
    Case de 25 (1UN)
    Case de 25 (1UN) - $1,192.82
    $1,192.82
    Each (1UN)
       $1,094.33
    Case de 25 (1UN)
       $1,192.82
  • Oven Thermometer
    +95...+115 °C
    200 mm
    0.1 °C
    ±0.4 °C
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,190.24
    $1,190.24
    Log in Para disponibilidad
    Case de 5 (1UN)
    Case de 5 (1UN) - $1,297.36
    $1,297.36
    Each (1UN)
       $1,190.24
    Case de 5 (1UN)
       $1,297.36
  • 89095-672
    Freezer Thermometer
    Freezer Thermometer
    −25...−5 °C
    210 mm
    0.1 °C
    ±0.4 °C
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,263.39
    $1,263.39
    Log in Para disponibilidad
    Case de 5 (1UN)
    Case de 5 (1UN) - $1,377.10
    $1,377.10
    Each (1UN)
       $1,263.39
    Case de 5 (1UN)
       $1,377.10