BRAND BRANDplates® pureGrade™ S Microplates, Sterile, BrandTech®
Catalog # 89093-608
Supplier: BRAND GMBH & CO. MX
Specifications
- Well Volume:330 µL
- Color:Transparent
- Descripción:96-Well Standard Microplates
- Referencia:89093-608
- Empaquetado:50 Individually Wrapped Plate/Lids
- Bottom Style:U-bottom
- Nº de proveedor:781660
Specifications
About this item
BRANDplates® pureGrade™ S plates are sterile, untreated plates designed for a variety of standardized applications
- Untreated, "medium binding" plates
- ANSI-SLAS formatted
- Sterility assurance level of 10-6
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. White and black 96-Well plates have a gray embossed alphanumeric code for quick and easy well identification.
Made of virgin polystyrene, these plates are suitable for homogenous assays, screening, and storage. Plates are sterilized by beta irradiation according to Ph. Eur. and USP 29, to a Sterility Assurace Level of 10-6.
Ordering information: All plates are supplied wth lids in packs of 50.