Orders LinkContactUsLinkComponent
Agar | Avantor
Agar
Highest Quality, Widest Selection for Every Application
Prepared in our on-site labs by experienced biologists and lab technicians, we have the widest selection of prepared media in a variety of packaging options to suit your needs. Available options vary by individual product.
Tubes
Set of twelve 16 x 125mm screw cap tubes. Slanted, unless noted.
Media Pour Pack
Package of 20 sterile Petri plates (90 x 15mm) and three media bottles. Each bottle pours seven plates.
Plates
Ten 90 x 15mm plates, sealed. Can be stored for up to two weeks.
Bottle
Six 125mm bottles, sufficient for 15-20 tubes or 5-7 plates per bottle.
Agar-Cubes-400x495.jpg
Pre-mixed, pre-poured, pre-measured and pre-cut cubes save you time and eliminate messy preparation in your lab.
Powdered-Agar-400x495.jpg
Fast-dissolving powders come premixed for quick preparation using neutral liquids.
Prepared-Agar-Plates-Tubes-And-Bottles-400x495.jpg
A wide selection for the scientific study of microorganisms. Ideal for educational and teaching laboratories.