Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
EDTA TETRASODICO 500G
Catalog # 85024-116
Supplier:
SPECTRUM CHEMICAL AND MX
New Product
Expand image
EDTA TETRASODICO 500G
Catalog # 85024-116
Supplier:
SPECTRUM CHEMICAL AND MX
Supplier Number:
E1047-500GM
New Product
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Target™ All-Plastic Disposable Luer-Slip Syringes, Thermo Scientific
Supplier: Thermo Scientific
HSW® Norm-Ject® Sterile Luer-Lock Syringes, Air-Tite
Supplier: AIR-TITE