Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TNTPLUS 874 TENSIOACTIVOS 0,1-4 MG/L
Catalog # 85023-506
Supplier:
REACTIVOS Y EQUIP SA DE CV MX
New Product
Expand image
TNTPLUS 874 TENSIOACTIVOS 0,1-4 MG/L
Catalog # 85023-506
Supplier:
REACTIVOS Y EQUIP SA DE CV MX
Supplier Number:
TNT874-LM
New Product
Frequently Bought Together
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Nitric acid 67 - 70%, ULTREX® II Ultrapure Reagent, Ultrapure for ICP, for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS MX