$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
ACIDO TRIFLUOROACETICO PARA HPLC
85022-848
:
SIGMA ALDRICH QUIMICA,S DE RL MX
ACIDO TRIFLUOROACETICO PARA HPLC
85022-848
:
SIGMA ALDRICH QUIMICA,S DE RL MX
:
T6508-10X10AMP
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International