$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
TRAJE DE PROECCION BIOCLEAN G
85022-328
:
ANSELL COMMERCIAL MEXICO SA DE MX
TRAJE DE PROECCION BIOCLEAN G
85022-328
:
ANSELL COMMERCIAL MEXICO SA DE MX
:
S-BDFC-L
75982-202_22-1849
75982-202_22-2437
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH