Skip to Header
Skip to Main Content
Skip to Footer
Avanzado
Avanzado
SDS
Certificados
Estructura química
Datos del fabricante
0
$0.00
Order Entry
Iniciar sesión
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TABLETOP ORBITAL SHAKER SOLARIS 4000 I
Catalog # 85022-146
Proveedor:
THERMO FISHER SCIENTIFIC MX
Expand image
TABLETOP ORBITAL SHAKER SOLARIS 4000 I
Catalog # 85022-146
Proveedor:
THERMO FISHER SCIENTIFIC MX
Supplier Number:
SK4001CPKG
Frequently Bought Together
VWR® Disposable Petri Dishes
Proveedor: VWR International
VWR® Nitrile Examination Gloves, Blue
Proveedor: VWR International
VWR® Light-Duty Tissue Wipers
Proveedor: VWR International
VWR® Universal Pipette Tips
Proveedor: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Proveedor: BDH