Order Entry
Mexico
Orders LinkContactUsLinkComponent
Value Plastics® Certified Sanitary Adapters, ADCF Polypropylene
Value Plastics® Certified Sanitary Adapters, ADCF Polypropylene
Sanitary to Hose Barb Adapter, 1-1/2" Tri-Clamp to ³/₈" ID Hose Barb, ADCF PP; 100/PK, ⅜", Sanitary to Barb, Straight, 1½", PP, 1½"
Catalog # MFLX31515-91BLK
Supplier:   VALUE PLASTICS TE
Supplier #:  SFMX670-6005100
Value Plastics® Certified Sanitary Adapters, ADCF Polypropylene
Catalog # MFLX31515-91BLK
Supplier:  VALUE PLASTICS TE
Value Plastics® Certified Sanitary Adapters, ADCF Polypropylene
Value Plastics® Certified Sanitary Adapters, ADCF Polypropylene
Sanitary to Hose Barb Adapter, 1-1/2" Tri-Clamp to ³/₈" ID Hose Barb, ADCF PP; 100/PK, ⅜", Sanitary to Barb, Straight, 1½", PP, 1½"
Price
$18,690.00
Log in for availability
Each (100UN)
Product Details & Documents
Certificate of compliance included!
  • Animal derivative component free
  • Meet USP Class IV requirements
  • Sterilizable vis gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow—and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI.
Delivery information: Includes certificate of compliance.
Documents
Specifications
  • Barb size/tubing int.Ø (port 2):
    ⅜"
  • Connection Type:
    Sanitary to Barb
  • Flow pattern:
    Straight
  • Sanitary clamp size (Port 1):
    1½"
  • Description:
    Sanitary to Hose Barb Adapter, 1-1/2" Tri-Clamp to ³/₈" ID Hose Barb, ADCF PP; 100/PK
  • Fitting Material:
    PP
  • Sanitary Gasket Size:
    1½"
  • Sterilisation:
    Ethylene oxide (ETO) or gamma
  • Cat. No.:
    MFLX31515-91BLK