$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
ON/OFF SWITCH FOR 07528-10
MFLX80022-69
:
VWR International
ON/OFF SWITCH FOR 07528-10
MFLX80022-69
:
VWR International
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Universal Pipette Tips
: VWR International