Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ingreso de pedido
Estado del pedido
Su cuenta
Carrito
Mexico
Ingreso de pedido
Estado del pedido
Su cuenta
Carrito
ContactUsLinkComponent
3/8 X1/2IN FEP TUBE W/TC END
Catalog # MFLX90253-89
Supplier:
Avantor
Expand image
3/8 X1/2IN FEP TUBE W/TC END
Catalog # MFLX90253-89
Supplier:
Avantor
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International