Order Entry
Mexico
ContactUsLinkComponent
AW-Lake FEM-03 Gear Flow Sensor/Transmitter Accessory, Flow Monitor
AW-Lake FEM-03 Gear Flow Sensor/Transmitter Accessory, Flow Monitor
Catalog # MFLX32928-75
Proveedor:  VWR International
undefined
AW-Lake FEM-03 Gear Flow Sensor/Transmitter Accessory, Flow Monitor
Catalog # MFLX32928-75
Proveedor:  VWR International
Acerca de este artículo
Easily control flow of your gear flow sensors.
  • Monitor both flow rate and total flow
Monitor both flow rate and flow from pulse output flowmeters in any engineering unit with a push of a button. Programmable, compact, and easy-to-use. Input square wave, sine, or sawtooth frequency. Back-up totalizer lock-out feature prevents accidental erasure. High-speed microprocessor ensures fast and accurate reporting — includes a 4 to 20 mA output for recording or datalogging. Model 32928-76 has two channels to monitor dual flows and display them separately, as a sum, as a difference, or as a ratio.
  • Output:
    None
  • Power:
    110 VAC
  • Display type:
    Digital
  • Description:
    Gear Flow Sensor/Transmitter Accessory, Flow Monitor
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Cat. no.:
    MFLX32928-75
Frequently Bought Together