Order Entry
Mexico
ContactUsLinkComponent
L-K Industries Class 1 Div 2 Heated Oil Centrifuge for Long Cone Tubes, 120 VAC
L-K Industries Class 1 Div 2 Heated Oil Centrifuge for Long Cone Tubes, 120 VAC
Catalog # MFLX17700-00
Supplier:  L-K INDUSTRIES MS
L-K Industries Class 1 Div 2 Heated Oil Centrifuge for Long Cone Tubes, 120 VAC
Catalog # MFLX17700-00
Supplier:  L-K INDUSTRIES MS
Supplier Number:  LB14-C120C
  • Rotor type:
  • Long cone tube type
  • Min. speed:
  • 200 rpm
  • Max. speed:
  • 2300 rpm
  • Max. RCF:
  • 1535 ×g
  • Timer range:
  • Preset at 10 minutes
  • Temperature control:
  • Analog with LCD display
  • Min. temperature:
  • Ambient °C (Ambient °F)
  • Max. temperature:
  • 100 °C (212 °F)
  • Power:
  • 120 VAC, 60 Hz
  • Tube capacity:
  • 100 ml
  • Warranty:
  • 1 year
  • Currently Viewing

Specifications

About this item

Safety first—Class 1, Division 2 certified models available.

  • Programmable digital controls maintain sample temperature and RPM accurately
  • Motor controller-integrated timer ensures test method compliance
  • Integral mounting flanges allow secure attachment to benchtop, stand, or table
  • Conformance: ASTM D91, D96, D893, D1796, D2273, D2709, and D4007
  • Shipping weight: 110 lb (50 kg)

These benchtop laboratory centrifuges help you meet the requirements of a variety of ASTM methods for oil testing. Program temperature and speed with digital precision, set motor control-integrated timer for test duration, and start the run. Simple controls save time, automated functions allow you to perform other tasks, and safety features protect the lab.

Choose explosion-proof models for hazardous location operation. Additional safety features include a lid-locking system preventing motor operation when open, emergency power-off palm button, and indicator lamps.

Order sample tubes separately.

Note: All models must be hard-wired by a qualified electrician with 12 AWG wire.

Delivery: Includes four-place rotor with buckets.
  • Max. speed:
    2300 rpm
  • Power:
    120 VAC, 60 Hz
  • Description:
    Class 1 Div 2 Heated Oil Centrifuge for Long Cone Tubes, 120 VAC
  • Environmentally Preferable:
  • Min. speed:
    200 rpm
  • Rotor type:
    Long cone tube type
  • Tube capacity:
    100 ml
  • Min. temperature:
    Ambient °C (Ambient °F)
  • Timer range:
    Preset at 10 minutes
  • Temperature control:
    Analog with LCD display
  • Max. RCF:
    1535 ×g
  • Max. temperature:
    100 °C (212 °F)
  • Warranty:
    1 year
  • Cat. no.:
    MFLX17700-00
  • Plug Type:
    No plug, hard-wiring required, Class 1 Div 2 approved
Frequently Bought Together