Order Entry
Mexico
Orders LinkContactUsLinkComponent
Electrically and Pneumatically Actuated Elliptical Valves
Electrically and Pneumatically Actuated Elliptical Valves
Three-Way Elliptical Valve, PP, Pneumatic, 3/4" NPT(F), PP, NPT(F), 0.75 NPT(F), ¾" NPT(F), ¾" NPT(F)
Catalog # MFLX98154-16
Electrically and Pneumatically Actuated Elliptical Valves
Catalog # MFLX98154-16
Supplier:  
Electrically and Pneumatically Actuated Elliptical Valves
Electrically and Pneumatically Actuated Elliptical Valves
Three-Way Elliptical Valve, PP, Pneumatic, 3/4" NPT(F), PP, NPT(F), 0.75 NPT(F), ¾" NPT(F), ¾" NPT(F)
Discontinued
Product Details & Documents
Completely disassemble in-line for easy cleaning.
  • FDA-compliant
  • Choose between Polypropylene (PP) or PVDF valve options
Use these actuated elliptical valves for liquid, gas, and vacuum applications. Valve systems are available in PP or PVDF bodies; all are FDA compliant and have O-rings made of Viton®. Rated for 150 psi at 73 °F (10 bar at 22 °C) (liquids).
Specifications
  • Connection Type:
    NPT(F)
  • Port Size:
    0.75 NPT(F)
  • Description:
    Three-Way Elliptical Valve, PP, Pneumatic, 3/4" NPT(F)
  • Inlet Port:
    ¾" NPT(F)
  • Outlet Port:
    ¾" NPT(F)
  • Housing Material:
    PP
  • Cat. No.:
    MFLX98154-16
  • Media compatibility:
    Use for liquid, gas, and vacuumapplications
  • Warranty:
    1 Month
  • Orifice:
    19.05 mm (0.75")
  • Actuation motor:
    Pneumatic