Order Entry
Mexico
ContactUsLinkComponent
Smart Gasket G-TH-150-S-4 1 1/2" Tri-Clamp, 4 Port, Silicone, Each
Smart Gasket G-TH-150-S-4 1 1/2" Tri-Clamp, 4 Port, Silicone, Each
Catalog # MFLX30545-12
Supplier:  VWR International
undefined
Smart Gasket G-TH-150-S-4 1 1/2" Tri-Clamp, 4 Port, Silicone, Each
Catalog # MFLX30545-12
Supplier:  VWR International
About this item
Smart Gasket lets you take samples or readings from a sanitary line.
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
  • Sanitary clamp size (Port 1):
    1½"
  • Connection type:
    Sanitary
  • Description:
    1 1/2" Tri-Clamp, 4 Port, Silicone, Each
  • Environmentally Preferable:
  • No. of ports:
    4
  • Warranty:
    1 year
  • Cat. no.:
    MFLX30545-12
  • Fitting Material:
    Platinum-cured silicone
  • Sanitary Gasket Size:
    1½"
Frequently Bought Together