Order Entry
Mexico
Orders LinkContactUsLinkComponent
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
1 1/2" Tri-Clamp, 3 Port, Viton, Each, 1½", Viton®, 1½"
Catalog # MFLX30544-14
Supplier:   VWR International
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Catalog # MFLX30544-14
Supplier:  VWR International
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
1 1/2" Tri-Clamp, 3 Port, Viton, Each, 1½", Viton®, 1½"
Price
$2,580.00
Log in for availability
Each (1UN)
Product Details & Documents
Smart Gasket lets you take samples or readings from a sanitary line.
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
Specifications
  • Sanitary clamp size (Port 1):
    1½"
  • Description:
    1 1/2" Tri-Clamp, 3 Port, Viton, Each
  • Fitting Material:
    Viton®
  • Sanitary Gasket Size:
    1½"
  • Cat. No.:
    MFLX30544-14
  • Warranty:
    1 year
  • Connection type:
    Sanitary
  • No. of ports:
    3