Order Entry
Mexico
ContactUsLinkComponent
Smart Gasket G-TH-200-V-1 2" Tri-Clamp Gasket, 1 Port, Viton, Each
Smart Gasket G-TH-200-V-1 2" Tri-Clamp Gasket, 1 Port, Viton, Each
Catalog # MFLX30542-18
Supplier:  VWR International
undefined
Smart Gasket G-TH-200-V-1 2" Tri-Clamp Gasket, 1 Port, Viton, Each
Catalog # MFLX30542-18
Supplier:  VWR International
About this item
Smart Gasket lets you take samples or readings from a sanitary line.
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
  • Sanitary clamp size (Port 1):
    2"
  • Connection type:
    Sanitary
  • Description:
    2" Tri-Clamp, 1 Port, Viton, Each
  • Environmentally Preferable:
  • No. of ports:
    1
  • Warranty:
    1 year
  • Cat. no.:
    MFLX30542-18
  • Fitting Material:
    Viton®
  • Sanitary Gasket Size:
    2"
Frequently Bought Together