Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TEE LUER F-M-F NYL 1000/PK
Catalog # MFLX30495-84
Supplier:
VWR International
Certificates
Expand image
TEE LUER F-M-F NYL 1000/PK
Catalog # MFLX30495-84
Supplier:
VWR International
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
MFLX45502-85_300758
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH