Order Entry
Mexico
ContactUsLinkComponent
Spirax Sarco IT 1-1/2" 60314  Y Steam Strainer, Cast Iron, 1-1/2" NPT(F)
Spirax Sarco IT 1-1/2" 60314  Y Steam Strainer, Cast Iron, 1-1/2" NPT(F)
Catalog # MFLX07600-78
Supplier:  VWR International
Spirax Sarco IT 1-1/2" 60314  Y Steam Strainer, Cast Iron, 1-1/2" NPT(F)
Catalog # MFLX07600-78
Supplier:  VWR International
About this item
Keep steam space free of condensate to provide maximum system efficiency.
  • Constructed of durable cast iron
Protect your pipeline from debris with these strainers.
  • Thread size NPT(F) (port 1):
    1.5"
  • Connection type:
    NPT(F)
  • Max. temperature:
    406 °F
  • Description:
    Cast iron "Y" strainer, 1-1/2" NPT(F)
  • Environmentally Preferable:
  • Max. operating pressure:
    300 psi
  • Warranty:
    1 year
  • Cat. no.:
    MFLX07600-78
  • Fitting Material:
    Cast iron
Frequently Bought Together