Order Entry
Mexico
ContactUsLinkComponent
 
Masterflex® Ismatec® Footswitch for Ecoline Drives, 8-Pin DIN Connector; 6-ft (1.8 m)
  MFLX78016-95
undefined
Masterflex® Ismatec® Footswitch for Ecoline Drives, 8-Pin DIN Connector; 6-ft (1.8 m)
  MFLX78016-95
 :  IS3572A

 

Simplify your dispensing.

  • Select the footswitch with connectio suitable for your Ismatec® pump

Footswitches provide convenient, hands-free operation of your pump drives from up to six feet away. Simply press once to start pump, press again to stop.

  • Description:
    Footswitch for Ecoline Drives, 8-Pin DIN Connector; 6-ft (1.8 m)
  • Length:
    183 cm (72")
  • Environmentally Preferable:
  • Model:
    78016-95
  • Input:
    8-Pin DIN
  • Warranty:
    1 year
  • Series:
    Ismatec Ecoline Drives
  • Cat. no.:
    MFLX78016-95
Frequently Bought Together