Order Entry
Mexico
Orders LinkContactUsLinkComponent
Replacement PFA Compression Ferrule Nut and Assemblies
Replacement PFA Compression Ferrule Nut and Assemblies
Ferrule plug, PFA, ¾", Compression Plug, PFA
Catalog # MFLX31320-49
CE certificate
Replacement PFA Compression Ferrule Nut and Assemblies
Catalog # MFLX31320-49
Supplier:  
CE certificate
Replacement PFA Compression Ferrule Nut and Assemblies
Replacement PFA Compression Ferrule Nut and Assemblies
Ferrule plug, PFA, ¾", Compression Plug, PFA
Discontinued
Product Details & Documents
Create a secure connection with compression fittings.
  • Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
  • Connection Type:
    Compression Plug
  • Description:
    Ferrule plug, PFA, ¾"
  • Fitting Material:
    PFA
  • Cat. No.:
    MFLX31320-49
  • Warranty:
    18 months
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)