Order Entry
Mexico
ContactUsLinkComponent
Masterflex® Fitting, PVDF, Tee, Hosebarb to Threaded Adapter, 1/4" ID x 1/4" ID x 3/8" NPT(M); 10/PK
Masterflex® Fitting, PVDF, Tee, Hosebarb to Threaded Adapter, 1/4" ID x 1/4" ID x 3/8" NPT(M); 10/PK
Catalog # MFLX40615-73
Supplier:  VWR International
undefined
Masterflex® Fitting, PVDF, Tee, Hosebarb to Threaded Adapter, 1/4" ID x 1/4" ID x 3/8" NPT(M); 10/PK
Catalog # MFLX40615-73
Supplier:  VWR International
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
  • Barb size/tubing int.Ø (port 1):
    0.25"
  • Barb size/tubing int.Ø (port 2):
    ¼"
  • Connection Type:
    Thread to Barb
  • Flow pattern:
    T
  • Description:
    Fitting, PVDF, Tee, Hosebarb to Threaded Adapter, 1/4" ID x 1/4" ID x 3/8" NPT(M); 10/PK
  • Environmentally Preferable:
  • Orifice dimensions:
    0.125"
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Cat. no.:
    MFLX40615-73
  • Fitting Material:
    PVDF
  • Barb Size / Tubing I.D. (Port 3):
    ¼"
Frequently Bought Together