Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Iniciar sesión
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PUMP MFLX I/P MODULAR US NEMA 230V
Catalog # MFLX77962-20-USN
Supplier:
VWR International
Ampliar imagen
PUMP MFLX I/P MODULAR US NEMA 230V
Catalog # MFLX77962-20-USN
Supplier:
VWR International
Related Information
Documentation
Manual
Manual Masterflex® IP® Series Precision Modular Drives
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International