Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PUMP MFLX LS EIP 600R EL2THN EU 230V
Catalog # MFLX77715-66-EU
Supplier:
VWR International
Expand image
PUMP MFLX LS EIP 600R EL2THN EU 230V
Catalog # MFLX77715-66-EU
Supplier:
VWR International
Related Information
Documentation
Manual
Manual Masterflex® L-S® Series Dispensing Pump Drives
Documentation
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International