$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
MODULO P/ HUMEDAD KBF/E5.3D, BINDER
85020-430
:
BINDER INC MX
MODULO P/ HUMEDAD KBF/E5.3D, BINDER
85020-430
:
BINDER INC MX
:
8009-1062
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International