Saint Gobain Sanitary Tubing Adapter; Polypropylene; 3/4" Mini to Female Luer Lock
Catalog # MFLX31200-50
Supplier: VWR International
CE certificate
Specifications
See all product configurationsSpecifications
About this item
Attach with just a twist!
- Choose between polypropylene (PP) and PVDF material options
Standard connection type for syringes, plus ideal for many applications with small tubing. These Female luer lock x 3/4" mini sanitary adapters are available in either polypropylene or PVDF.
- Connection Type:Sanitary to Female Luer
- Flow pattern:Straight
- Sanitary clamp size (Port 1):¾"
- Description:Tubing Adapter; Polypropylene (PP), ¾" mini to female luer lock
- Environmentally Preferable:
- Warranty:None
- Cat. no.:MFLX31200-50
- Fitting Material:PP
Frequently Bought Together