Gauge Guard Isolator Seal, EPDM; 1-1/2" Tri-Clamp
Catalog # MFLX30565-19
Supplier: VWR International
3A Sanitary approval
About this item
Extend the life and accuracy of your expensive and fragile instruments.
- Manufactured to meet USP Class VI certification
- Animal derived ingredient free
Gauge Guard Protectors safeguard your expensive instruments from damaging process fluids and destructive routine maintenance such as autoclaving. Gauge guard is platinum-cured silicone combined with a 316 stainless steel perforated plate, and slips over the OD of your diaphragm seal. Model 30565-03 fits both 1 and 1 1/2" seals.
Gauge Guard Isolator Seals combine a hygenic seal with a membrane to isolate hot or caustic solutions from your instrument. Available in platinum-cured silicone or EPDM.
- Description:Gauge Guard Isolator Seal, EPDM; 1-1/2" Tri-Clamp
- Environmentally Preferable:
- Warranty:1 year
- Cat. no.:MFLX30565-19
Frequently Bought Together