Order Entry
Mexico
Orders LinkContactUsLinkComponent
PendoTECH® Single-Use Pressure Sensor Accessories
PendoTECH® Single-Use Pressure Sensor Accessories
Adapter Tee, 1/2" Hose Barb x 1/2" Hose Barb x Male Luer Fitting, Polycarbonate
Catalog # MFLX19406-35
CE Compliant
Supplier:   PENDOTECH
Supplier #:  PDKT-105-03
PendoTECH® Single-Use Pressure Sensor Accessories
Catalog # MFLX19406-35
Supplier:  PENDOTECH
CE Compliant
PendoTECH® Single-Use Pressure Sensor Accessories
PendoTECH® Single-Use Pressure Sensor Accessories
Adapter Tee, 1/2" Hose Barb x 1/2" Hose Barb x Male Luer Fitting, Polycarbonate
Price
$570.00
Log in for availability
Each (1UN)
Product Details & Documents
Optimize performance of your PendoTECH® Pressure Sensor.
  • Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Documents
Specifications
  • Description:
    Adapter Tee, 1/2" Hose Barb x 1/2" Hose Barb x Male Luer Fitting, Polycarbonate
  • Cat. No.:
    MFLX19406-35
  • Max. pressure:
    76 psi
  • Warranty:
    3 Months
  • Process connection:
    1/2" x 1/2" hose barb, male luer