$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
FINGER SNAP LUER LK RG,RED NLN,PK OF 100
Catalog # MFLX15952-61
Supplier:
VWR International
Expand image
FINGER SNAP LUER LK RG,RED NLN,PK OF 100
Catalog # MFLX15952-61
Supplier:
VWR International
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International