$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
FINGER SNAP LUER LK RG,ORG NLN PK OF 100
Catalog # MFLX15952-43
Proveedor:
VWR International
Expand image
FINGER SNAP LUER LK RG,ORG NLN PK OF 100
Catalog # MFLX15952-43
Proveedor:
VWR International
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Proveedor: VWR International
VWR® Universal Pipette Tips
Proveedor: VWR International
VWR® Nitrile Examination Gloves, Blue
Proveedor: VWR International
VWR® Disposable Petri Dishes
Proveedor: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Proveedor: BDH