$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
FLOW METER 1.25IN. 10/100 GPM
MFLX00336-UM
:
BADGER METER INC
FLOW METER 1.25IN. 10/100 GPM
MFLX00336-UM
:
BADGER METER INC
:
H812A-100
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International