Masterflex® I/P® Replacement Main Body, PPS Housing, SS Rotor
Catalog # MFLX77602-17
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® I/P® Easy-Load® Pump Head with these replacement parts.
- Suitable for use with pump heads 77601-60 and 77601-62
Keep your pump head operating at its best with these replacement main body and occusion bed assemblies.
- Pump series:I/P
- Description:Replacement Main Body, PPS Housing, SS Rotor
- Environmentally Preferable:
- Warranty:3 Months
- Cat. no.:MFLX77602-17
- Housing Material:PPS
Frequently Bought Together