Masterflex® L/S® Gear Service Kits for 10 to 600 rpm Digital Modular Drives
MFLX77300-01
Make the most of your Masterflex drives replacement parts and service kits.
- Easy to install, these service kits avoid costly downtime of your gear pump head
- Suitable for use with drives 77301-20 and -30.
Extend the life of your Masterflex pump with these replacement parts.
: Includes gear assembly (nylon gear/shaft), and gear case cover gasket.
- Description:Gear Service Kit for Modular Drives 77301-20/-30
- Environmentally Preferable:
- Warranty:2 years
- Compatible with:77301-20 or 77301-30
- Cat. no.:MFLX77300-01
Frequently Bought Together