Masterflex® Drum Pump Inlet Tube, CPVC, 47" L; 1/Each
Catalog # MFLX70604-68
Supplier: VWR International
About this item
Safely transfer corrosive chemicals, flammable liquids, and non-corrosive media.
- Heavy-duty, thick-walled construction makes tube more durable
- Tube length 47"
Replacement tubes do not include barrel adapter and mouting bracket; order separately.
- Description:Drum Pump Tube Set, CPVC, 47"L
- Environmentally Preferable:
- Model:70604-68
- Wetted materials:CPVC
- Tube length, inlet:47"
- Warranty:3 years
- Cat. no.:MFLX70604-68
Frequently Bought Together