Masterflex® Replacement Drum Pump Tubes, Avantor®
Drum Pump Tube Set, PP, 39" L
Catalog # MFLX70604-42
Supplier: VWR International
Masterflex® Replacement Drum Pump Tubes, Avantor®
Catalog # MFLX70604-42
Supplier: VWR International
Jump to:
Product Details & Documents
Safely transfer corrosive chemicals, flammable liquids, and non-corrosive media.
- Heavy-duty, thick-walled construction makes tube more durable
Replacement tubes do not include barrel adapter and mouting bracket; order separately.
Specifications
- Description:Drum Pump Tube Set, PP, 39" L
- Cat. No.:MFLX70604-42
- Warranty:4 months
- Tube length, inlet:39"
- Model:70604-42
- Wetted materials:Polypropylene