Order Entry
Mexico
ContactUsLinkComponent
Compression pipe adapter, Brass, 5/8"×1/2", Each
Compression pipe adapter, Brass, 5/8"×1/2", Each
Catalog # MFLX31412-78
undefined
Compression pipe adapter, Brass, 5/8"×1/2", Each
Catalog # MFLX31412-78
Supplier Number:  IBD10MC8
Acerca de este artículo
Create a secure connection with compression fittings.
  • Constructed of durable brass
Good chemical resistance. Rated to 400 psi maximum at 77 °F ( 25 °C). Usable from -85 to 482 °F (-65 to 250 °C).
  • Connection Type:
    Compression to Thread
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 2):
    ½"
  • Description:
    Compression pipe adapter, Brass, 5/8"×1/2", Each
  • Environmentally Preferable:
  • Warranty:
    LTD life time
  • Cat. no.:
    MFLX31412-78
  • Fitting Material:
    Brass
Frequently Bought Together