Compression Elbow Threaded Adapter, Stainless Steel, 1/4"×1/4", Each
Catalog # MFLX31406-78
Proveedor: MIDWEST VALVE & FITTING INC
CE certificate
Acerca de este artículo
Create a secure connection with compression fittings.
- Constructed of durable stainless steel (SS)
Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC).
- Connection Type:Compression to Thread
- Flow pattern:Elbow
- Thread size NPT(M) (port 2):¼"
- Description:Compression Elbow Threaded Adapter, Stainless Steel, 1/4"×1/4", Each
- Environmentally Preferable:
- Sterilisation:Autoclave, ethylene oxide (ETO), or gamma
- Warranty:LTD life time
- Cat. no.:MFLX31406-78
- Fitting Material:Stainless steel 316
Frequently Bought Together